di-Natriumtetraborat Decahydrat 99,0-105,0% NF, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

7418-26EA 0 EUR
7418-26
di-Natriumtetraborat Decahydrat 99,0-105,0% NF, Macron Fine Chemicals™
di-Natriumtetraborat Decahydrat
Formel: Na₂B₄O₇·10H₂O
Siedepunkt: 320 °C (1013 hPa)
Schmelzpunkt: 75 °C
Dichte: 1,73 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149193
CAS-Nummer: 1303-96-4
EINECS: 215-540-4
REACH: 01-2119490790-32
Merck Index: 13,08662

Bestellen

Spezifikation Testergebnisse

Meets N.F. Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical 99.0 - 105.0 %
NF - Assay (Na₂B₄O₇·10H₂O) 99.0 - 105.0 %
NF - Carbonate and Bicarbonate Passes Test
NF - Identification A Passes Test
NF - Identification B Passes Test
Appearance (colorless crystals or white crystalline powder)
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR