Natriumhydrogencarbonat 99,0-100,5% (Trockenmasse), Pulver USP, ACS, J.T.Baker®

Lieferant: Avantor

7451-07EA 0 EUR
7451-07 7451-26
Natriumhydrogencarbonat 99,0-100,5% (Trockenmasse), Pulver USP, ACS, J.T.Baker®
Natriumhydrogencarbonat

Bitte beachten This product is not intended for use in hemodialysis.

Formel: NaHCO₃
Dichte: 2,20 g/cm³
MDL: MFCD00003528
CAS-Nummer: 144-55-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets U.S.P Requirements
Meets ACS Reagent Chemical Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical = 5 ppm
ACS - Ammonium (NH₄) ≤5 ppm
ACS - Assay (NaHCO₃) (by acidimetry) 99.7 - 100.3 %
ACS - Calcium (Ca) ≤0.02 %
ACS - Chloride (Cl) ≤0.003 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Insoluble Matter ≤0.015 %
ACS - Iron (Fe) ≤0.001 %
ACS - Magnesium (Mg) ≤0.005 %
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Potassium (K) ≤0.005 %
ACS - Sulfur Compounds (as SO₄) ≤0.003 %
USP - Assay (NaHCO₃) (dried basis) 99.0 - 100.5 %
USP - Chloride (Cl) ≤0.015 %
USP - Identification Passes Test
USP - Insoluble Substances Passes Test
USP - Limit of Ammonia ≤20 ppm
USP - Limit of Sulfur Compounds ≤0.015 %
USP - Loss on Drying ≤0.25 %
USP - Normal Carbonate Passes Test
USP - Arsenic (As) ≤2 ppm
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
This product is not intended for use in hemodialysis.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR