Natriumchlorid, wasserfrei ≥99% ACS, frei fließend, Fluka™

Lieferant: Honeywell Chemicals

Synonyme: Saline, NaCl, Sea salt, Halite

746398-2.5KG 746398-4X2.5KG 746398-500G 746398-6X500G
746398-2.5KGEA 126.56 EUR
746398-2.5KG 746398-4X2.5KG 746398-500G 746398-6X500G
Natriumchlorid, wasserfrei ≥99% ACS, frei fließend, Fluka™
Natriumchlorid

Free flowing salt.

Formel: NaCl
Molecular Weight: 58.44 g/mol
Siedepunkt: 1.465 °C (1.013 hPa)
Schmelzpunkt: 800 °C
Dichte: 2.160 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003477
CAS-Nummer: 7647-14-5
Merck Index: 14,08599

Bestellen

Spezifikation Testergebnisse

Assay 99.0 - 101.5 %
Turbidity of solution complying
Colour of the solution complying
Flowability complying
pH (5 %, 25 °C) 5.0 - 9.0
Water insoluble matter Max. 0.005 %
Barium (Ba) complying
Calcium (Ca) Max. 0.002 %
Iron (Fe) Max. 2 ppm
Potassium (K) Max. 0.005 %
Magnesium (Mg) Max. 0.001 %
Heavy metals (ICP) ACS Max. 5 ppm
Bromide (Br) Max. 0.01 %
Chlorate, Nitrate (as NO₃) Max. 0.003 %
Iodide (I) Max. 0.002 %
Phosphate (PO₄) Max. 5 ppm
Sulfate (SO₄) Max. 0.004 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR