n-Octan ≥99,0% (durch GC), Puriss. p.a., Riedel-de Haën™

Lieferant: Honeywell Chemicals
Gefahr

74821-500ML
74821-500MLEA 0 EUR
74821-500ML
n-Octan ≥99,0% (durch GC), Puriss. p.a., Riedel-de Haën™
n-Octan

Beilstein Number: 1696875
RTECS Number: RG8400000
UNSPSC Code: 12190000

Formel: H₃C(CH₂)₆CH₃
Molecular Weight: 114.23 g/mol
Siedepunkt: 6.5 %(V)
Schmelzpunkt: –57 °C
Dichte: 0.703 g/cm³ (25 °C)
Flammpunkt: 13 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009556
CAS-Nummer: 111-65-9
EINECS: 203-892-1
UN: 1262
ADR: 3 II
Merck Index: 13,06782
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68adf890d68728825d4a2fa4b032700b93f710b8&vid=b00a2925902bc425c80f8ebb3f0e7b456c800ae3&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Identity (NMR) complying
Assay (GC) Min. 99.0 %
Refractive index (n 20/D) 1.397 - 1.399
Non-volatile matter Max. 0.002 %
Water (Karl Fischer) Max. 0.02 %
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
Explosion Limit 6.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR