Lead standard for ICP-MS, TraceCERT®, Supelco®
Lieferant: Merck
75015-100ML
75015-100MLEA
240
EUR
75015-100ML
Lead standard for ICP-MS, TraceCERT®, Supelco®
Standards
Kationischer Standards
Blei-Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034