Organic reference standard, 2,3-Hexanedione, analytical standard, Supelco®
Lieferant: Merck
75034-100MG
75034-100MGEA 125 EUR
75034-100MG
Organic reference standard, 2,3-Hexanedione, analytical standard, Supelco®
Standards Organische Standards
2,3-Hexanedione can be used as an internal standard for the quantitative determination of methylglyoxal present in the tissues of Sprague-Dawley rats using electrospray ionization liquid chromatography coupled with mass spectrometry(ESI/LC/MS).
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0b91f4105059ab6cab7c658f6c15ba2ec7bfd6b&vid=f0b91f4105059ab6cab7c658f6c15ba2ec7bfd6b&prid=web&clid=SAPDAM