Organic reference standard, ethyl vanillin, analytical standard, Supelco®
Lieferant: Merck
75042-100MG
75042-100MGEA 51.2 EUR
75042-100MG
Organic reference standard, ethyl vanillin, analytical standard, Supelco®
Standards Organische Standards
Ethyl vanillin is used as a reference standard in the quantitative determination of its presence in vanillin extract products using headspace-solid phase microextraction (HS-SPME) combined with gas chromatography coupled with mass spectrometry (GC–MS) and liquid chromatography coupled with mass spectrometry (LC-MS).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aea06749de3399f9b3dc3290ee839f0662c28ecc&vid=1c5030c506ac14057e431da0b292fa4c9c450b42&prid=web&clid=SAPDAM