Organic reference standard, ethyl vanillin, analytical standard, Supelco®
Lieferant: Merck
75042-100MG
75042-100MGEA
51.2
EUR
75042-100MG
Organic reference standard, ethyl vanillin, analytical standard, Supelco®
Standards
Organische Standards
Ethyl vanillin is used as a reference standard in the quantitative determination of its presence in vanillin extract products using headspace-solid phase microextraction (HS-SPME) combined with gas chromatography coupled with mass spectrometry (GC–MS) and liquid chromatography coupled with mass spectrometry (LC-MS).