1,2-Benzisothiazol-3(2H)-on, Supelco®

Lieferant: Merck

Synonyme: 1,2-Benzisothiazol-3-one

MQ100

75169-100MG
75169-100MGEA 169 EUR
75169-100MG
1,2-Benzisothiazol-3(2H)-on, Supelco®
1,2-Benzisothiazol-3(2H)-on
Formel: C₇H₅NOS
Molecular Weight: 151.19 g/mol
MDL: MFCD00044001
CAS-Nummer: 2634-33-5
EINECS: 220-120-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=910a034c7eb313f4022e0195c357e255e6a1e739&vid=23536031edb19c7f3f97de7e10e2b0cf27cc6b28&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR