Gram Entfärbelösung zur Gramfärbung, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

75482-2.5L-F 75482-250ML-F
75482-2.5L-FEA 106 EUR
75482-2.5L-F 75482-250ML-F
Gram Entfärbelösung zur Gramfärbung, Sigma-Aldrich®
Gram Entfärbelösung

Gram staining is one of the most significant staining techniques in microbiology. Gram′s decolouriser solution is a mixture of ethanol and acetone. Gram′s decolouriser solution is used in Gram staining procedure, after the iodine step to remove the crystal violet dye. The decolouriser dissolves the lipid layer from the gram-negative cells.

Dichte: 0,80 g/ml at 20 °C
Flammpunkt: 20 °C
Lagertemperatur: Room Temperature
MDL: MFCD00240361
CAS-Nummer: 37348-17-7

Bestellen

Spezifikation Testergebnisse

Appearance (colour) Colourless
Appearance (form) Liquid
Density D20/4 0.785 - 0.814
Refractive index N20/D 1.356 - 1.366
Proton NMR spectrum Conforms to structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR