Osmiumtetraoxid -Lösung, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
75633-2ML
75633-2MLEA
154
EUR
75633-2ML
Osmiumtetraoxid -Lösung, Sigma-Aldrich®
Osmiumtetraoxid
Osmium tetroxide solution has been used as a fixative in electron microscopy.
Formel:
O₄Os Dichte: 1,0 g/mL (20 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00011150 CAS-Nummer: 20816-12-0 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...