Supelco® TraceCERT® Organic Reference Standard, 2,6-Di-tert-butyl-4-methylphenol
Lieferant: Merck
75918-50MG
75918-50MGEA 85 EUR
75918-50MG
Supelco® TraceCERT® Organic Reference Standard, 2,6-Di-tert-butyl-4-methylphenol
Standards Organische Standards
  • GHS signal: GHS09
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P273 - P391 - P501
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b2d64027cfe8421bf6d86b4ae67078bd6866857&vid=5b2d64027cfe8421bf6d86b4ae67078bd6866857&prid=web&clid=SAPDAM