Organic reference standard, Dimethyl sulfide, analytical standard, Supelco®
Lieferant: Merck
76048-100MG
76048-100MGEA
125
EUR
76048-100MG
Organic reference standard, Dimethyl sulfide, analytical standard, Supelco®
Standards
Organische Standards
Dimethyl sulfide is found to be the most abundant volatile thioether, which can find applications as a flavor enhancing agent in food. It can undergo oxidation in the process of malting and kilning, to yield dimethyl sulfoxide (DMSO).
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.