Total organic carbon standards, according to ISO/CEN EN1484, Supelco®
Lieferant: Merck
76067-250ML-F
76067-500ML-F
76067-250ML-FEA
114
EUR
76067-250ML-F
76067-500ML-F
Total organic carbon standards, according to ISO/CEN EN1484, Supelco®
Standards
Umweltanalyse-Standards
- Stabilized
- Limited shelf life, expiry date on the label
Bitte beachten Storage temperature: 2…8 °C