Orbital molecular models
Lieferant: SPIRING ENTERPRISES
MOS-900-4
761-0114EA 39.2 EUR
761-0114
Orbital molecular models
Modelle Molekularbiologie
Four models of molecular orbital configurations using benzene, ethane, ethene, and ethyne.
The models depict sigma and pi bonds and effects such as hybridisation and delocalised electrons.