Paraplast Plus®, Plätzchen für die Elektronenmikroskopie (fast-melting), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY


M-Clarity™ Qualitätsstufe = MQ100

76258-1KG
76258-1KGEA 44.3 EUR
76258-1KG
Paraplast Plus®, Plätzchen für die Elektronenmikroskopie (fast-melting), Sigma-Aldrich®
Paraffinwachs versetzt mit synthetischen Polymeren

Regular Paraplast with approx. 0,8% DMSO added for enhanced ease of infiltration and sectioning. Paraffin polyisobutylene mixture is a tissue embedding medium composed of highly purified paraffin and polyisobutylene and a low level of dimethyl sulfoxide (DMSO) to enhance the rate of tissue penetration. Paraffin-polyisobutylene (76258) supports thin wrinkle-free sectioning down to 4 µm with more flexible and continuous ribbons.


Paraffin – polyisobutylene mixture

Schmelzpunkt: 56,..57 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00131795
CAS-Nummer: 125387-89-5
EINECS: 232-315-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR