Ready-to-Load™ kits, whose DNA was left behind?
Lieferant: EDVOTEK
S-51
763-0008EA 71.5 EUR
763-0008
Ready-to-Load™ kits, whose DNA was left behind?
Lehrmittel Forensik-Lehrmittel Forensische Kits
DNA obtained from a single hair left behind at a crime scene can be used to identify a criminal. In this experiment, students will compare simulated crime scene DNA with that of 2 suspects.
  • For 10 lab groups
  • Complete in 45 minutes
Also required: Electrophoresis tank and power supply.
Lieferumfang: Kit includes instructions, Ready-to-Load™ dye sample, agarose powder, practice gel loading solution, electrophoresis buffer, and microtipped transfer pipettes.