Dicotyledon plant stem
Lieferant: Binhold, Paul Lehrmittel
1002506
763-0138EA 627 EUR
763-0138
Dicotyledon plant stem
Modelle Botanik
Phaseolus vulgaris. Model of dicotyledonous stem (beans) magnified 250X.
Stem cell types are shown longitudinally and in cross section.