Supelco® TraceCERT® Organic Reference Standard, Resveratrol

Lieferant: Merck

76511-100MG
76511-100MGEA 134 EUR
76511-100MG
Supelco® TraceCERT® Organic Reference Standard, Resveratrol
Standards Organische Standards

Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.


Bitte beachten P264 - P280 - P305 + P351 + P338 - P337 + P313

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=423db0939882d76b0dae0c1b964d10a9c0687a88&vid=423db0939882d76b0dae0c1b964d10a9c0687a88&prid=web&clid=SAPDAM

Bestellen

TECHNISCHE DATEN

Synonym(s) 3,4′,5-Trihydroxy-trans-stilbene; 5-[(1E)-2-(4-Hydroxyphenyl)ethenyl]-1,3-benzenediol
M-Clarity Qualitätsstufe MQ300
CAS Nr. 501-36-0
MDL No. MFCD00133799
Molecular weight 228,24
Beschreibung Gefahrenklasse H319
Shipping conditions STD
Lagerbedingungen 2…8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR