Organic reference standard, Urethane, analytical standard, Supelco®
Lieferant: Merck
76607-100MG
76607-100MGEA 130 EUR
76607-100MG
Organic reference standard, Urethane, analytical standard, Supelco®
Standards Organische Standards
Urethane is used as an anesthetic in animals, since it shows minimal effects on cardiovascular and respiratory systems and maintenance of spinal reflexes.
It can also be used in the synthesis of poly(urethane urea) segmented block, which can find applications as blood sacs in ventricular assist devices and total artificial hearts.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a4faa043d152999f28b379a8d7bb78395bf4bfd&vid=2a4faa043d152999f28b379a8d7bb78395bf4bfd&prid=web&clid=SAPDAM