Anthranilamid ≥99,0% (durch HPLC) Matrixsubstanz für MALDI-MS, Supelco®

Lieferant: Merck

Synonyme: 2-Aminobenzamid, Anthranilsäureamid, 2-Aminobenzoesäureamid

76884-1G
76884-1GEA 232 EUR
76884-1G
Anthranilamid ≥99,0% (durch HPLC) Matrixsubstanz für MALDI-MS, Supelco®
Anthranilamid

Used for non-selective, efficient fluorescent labeling of glycans. Slightly less sensitive than anthranilic acid (2-AA) for glycan labeling.

Formel: C₇H₈N₂O
Molecular Weight: 136,15 g/mol
MDL: MFCD00007981
CAS-Nummer: 88-68-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b7819b52f29c6db29e43451f2e63a405226372a&vid=6b7819b52f29c6db29e43451f2e63a405226372a&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Color) White
Appearance (Form) Powder or Crystals or Flakes
Titration (NT) HCLO4 0.1M 99 - 101 %
Purity (HPLC area %) ≥ 99.0 %
Proton NMR Spectrum Conforms to structure
Metal trace Analysis (ICP) Corresponds to requirements
BARIUM (ICP) ≤ 5.0 mg/kg
CALCIUM (ICP) ≤ 5.0 mg/kg
CADMIUM (ICP) ≤ 5.0 mg/kg
COBALT (ICP) ≤ 5.0 mg/kg
CHROMIUM (ICP) ≤ 5.0 mg/kg
COPPER (ICP) ≤ 5.0 mg/kg
IRON (ICP) ≤ 5.0 mg/kg
POTASSIUM (ICP) ≤ 50 mg/kg
MAGNESIUM (ICP) ≤ 5.0 mg/kg
MANGANESE (ICP) ≤ 5.0 mg/kg
SODIUM (ICP) ≤ 150 mg/kg
NICKEL (ICP) ≤ 5.0 mg/kg
LEAD (ICP) ≤ 5.0 mg/kg
ZINC (ICP) ≤ 5.0 mg/kg
RECOMMENDED RETEST PERIOD 48 MONTHS

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR