Palladium standard for ICP, TraceCERT®, Supelco®
Lieferant: Merck
77091-100ML
77091-100MLEA
174
EUR
77091-100ML
Palladium standard for ICP, TraceCERT®, Supelco®
Standards
Kationischer Standards
Palladium-Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034