Titan(II)oxid ≥99,5% (Metall-Basis)

Lieferant: Thermo Fisher Scientific

77126.06 77126.14 77126.22 77126.36
77126.06EA 42.3 EUR
77126.06 77126.14 77126.22 77126.36
Titan(II)oxid ≥99,5% (Metall-Basis)
Titan(II)oxid

Purity: 99.5% CAS No.: 12137-20-1 Molecular Formula: TiO Formula Weight: 63.90 Physical Form: -325 Mesh Powder Density: 4.95 Melting Point: 1700° Boiling Point: 3000° MDL No.: MFCD00036279

Formel: OTi
Siedepunkt: >3000 °C
Schmelzpunkt: 1700 °C
Dichte: 4,95
Lagertemperatur: Raumtemperatur
MDL: MFCD00036279
CAS-Nummer: 12137-20-1
EINECS: 235-236-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR