Titan(IV)ethanolat (≥33% TiO₂)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
CAS No.: 3087-36-3
Molecular Formula: C8H20O4Ti
Formula Weight: 228.15
Flash Point: 28°(84°F)
Physical Form: Liquid
Density: 1.088
Refractive Index: 1.5043
Sensitivity: Moisture Sensitive
Appearance: Pale yellow
Odour: Alcohol-like
Melting Point: -40°
Boiling Point: 150-152°/10mm
Solubility: Soluble in organic solvents. Decomposes in water
UN No.: 2924
MDL No.: MFCD00009071
Formel:
C₈H₂₀O₄Ti Siedepunkt: 150…152 °C (10 mmHg) Schmelzpunkt: –40 °C Dichte: 1,088 Flammpunkt: 28 °C (84 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009071 CAS-Nummer: 3087-36-3 EINECS: 221-410-8 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...