Ammoniumsulfat ≥99,0%, GenAR® für die Biotechnologie, Macron Fine Chemicals™
Lieferant: Avantor Performance Materials
Synonyme:
Diammoniumsulfat, di-Ammoniumsulfat
7725-88EA
441.57
EUR
7725-88
7725-22
7725-65
Ammoniumsulfat ≥99,0%, GenAR® für die Biotechnologie, Macron Fine Chemicals™
Ammoniumsulfat
Formel:
(NH₄)₂SO₄ Molecular Weight: 132.14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003391 CAS-Nummer: 7783-20-2 EINECS: 231-984-1 Merck Index: 13,00559 |
Spezifikation Testergebnisse
For Laboratory,Research,or Manufacturing Use | — |
ACS - Assay | ≥99.0 % |
Solubility | Passes Test |
ACS - Arsenic (As) | ≤0.00002 % |
ACS - Chloride (Cl) | ≤0.0002 % |
ACS - Heavy Metals (as Pb) | ≤0.0005 % |
ACS - Insoluble Matter | ≤0.005 % |
ACS - Iron (Fe) | ≤0.0003 % |
ACS - Nitrate (NO₃) | ≤0.0002 % |
ACS - Phosphate (PO₄) | ≤0.0005 % |
ACS - Residue on Ignition | ≤0.005 % |
ACS - pH of 5% Solution at 25°C | 5.0 - 6.0 |
Copper (Cu) | ≤1 ppm |
Zinc (Zn) | ≤1.000 ppm |
Enzyme Activity - DNase Activity | Passes Test |
Enzyme Activity - RNase Activity | Passes Test |
Enzyme Activity - Protease Activity | Passes Test |
Endotoxin Concentration (EU/g) | ≤2.5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...