EDTA Dinatriumsalz Dihydrat 99,0-101,0% (Trockenmasse), GenAR® USP für die Biotechnologie, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

7727-88EA 2042.4 EUR
7727-88 7727-04
EDTA Dinatriumsalz Dihydrat 99,0-101,0% (Trockenmasse), GenAR® USP für die Biotechnologie, Macron Fine Chemicals™
EDTA Dinatriumsalz Dihydrat
Formel: C₁₀H₁₄N₂Na₂O₈·2H₂O
Schmelzpunkt: 255 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00150037
CAS-Nummer: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Food GMP Manufactured Product
Meets U.S.P Requirements
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
USP - Assay (dried basis) 99.0 - 101.0 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Identification C Passes Test
USP - pH (1 in 20) 4.0 - 5.5
USP - Loss on Drying (by wt) 8.7 - 11.4 %
USP - Calcium (Ca) Passes Test
USP - Limit of Nitrilotriacetic Acid (by wt) ≤0.1 %
EP/BP - Appearance of Solution Passes Test
EP/BP - Identification A Passes Test
EP/BP - Assay 98.5 - 101.0 %
EP/BP - Identification B Passes Test
EP/BP - Identification D Passes Test
EP/BP - Impurity A and Related Substances - Impurity A ≤0.1 %
EP/BP - Iron (Fe) ≤80 ppm
EP/BP - pH (1 in 20) 4.0 - 5.5
Endotoxin Concentration (EU/g) ≤5
FCC - Assay (anhydrous basis) 99.0 - 101.0 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Identification C Passes Test
FCC - Lead (Pb) ≤10 mg/kg
FCC - pH (1 in 100) 4.3 - 4.7
FCC - Calcium (Ca) Passes Test
FCC - Limit of Nitrilotriacetic Acid (by wt) ≤0.1 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR