(-)-Perillyl alcohol 98, Supelco®

Lieferant: Merck

Synonyme: (S)-4-Isopropenyl-1-cyclohexenylmethanol, (S)-p-Mentha-1,8-dien-7-ol, (S)-(4-(Prop-1-en-2-yl)cyclohex-1-en-1-yl)methanol, (S)-(−)-Perillyl alcohol

MQ100

77311-1ML
77311-1MLEA 82 EUR
77311-1ML
(-)-Perillyl alcohol 98, Supelco®
(-)-Perillyl alcohol
Formel: C₁₀H₁₆O
Molecular Weight: 152.24 g/mol
MDL: MFCD00062995
CAS-Nummer: 18457-55-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c9d1dc5ec3dfec5ab8ba969156d00c39af21e2e&vid=a508ba4ed62629b8a4eb0afa14fab5be4adc1111&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR