Petroleumbenzin, 30…40 °C, Puriss. p.a. analytical reagent, hydrogen treated naphtha_low boiling pt, Riedel-de Haën™

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Benzin, Petrolether

77399-2.5L 77399-1L 77399-25L 77399-7L
77399-2.5LEA 180.92 EUR
77399-2.5L 77399-1L 77399-25L HONC77399-7L
Petroleumbenzin, 30…40 °C, Puriss. p.a. analytical reagent, hydrogen treated naphtha_low boiling pt, Riedel-de Haën™
Petroleumbenzin

UNSPSC Code: 12191502

Siedepunkt: 7.8 %(V)
Dichte: 0.628 g/cm³ (15 °C)
Flammpunkt: –56 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00081849
CAS-Nummer: 101316-46-5
EINECS: 309-852-0
UN: 1268
ADR: 3 I

Bestellen

Spezifikation Testergebnisse

Boiling range (30-40°C) Min. 95 %
Boiling range 30 - 40 °C
Density (D 20/4) 0.624 - 0.627
Refractive index (n 20/D) 1.356 - 1.358
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.02 %
Free acid (as CH₃COOH) Max. 0.005 %
Aluminium (Al) Max. 0.00005 %
Barium (Ba) Max. 0.00001 %
Bismuth (Bi) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00001 %
Potassium (K) Max. 0.00005 %
Lithium (Li) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Molybdenum (Mo) Max. 0.00001 %
Sodium (Na) Max. 0.00005 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.00001 %
Strontium (Sr) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
Iodine value Max. 2
Reaction against H₂SO₄ complying
Benzene (GC) Max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR