Kaliumdihydrogenphosphat 98,0-100,5% (Trockenmasse, RT), Kristalle, GenAR® NF, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Achtung

7746-88EA 1097.17 EUR
7746-88 7746-04
Kaliumdihydrogenphosphat 98,0-100,5% (Trockenmasse, RT), Kristalle, GenAR® NF, Macron Fine Chemicals™
Kaliumdihydrogenphosphat
Formel: KH₂PO₄
Siedepunkt: ∼100 °C (1013 hPa)
Schmelzpunkt: 253 °C
Dichte: 2,3 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011401
CAS-Nummer: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 13,07744

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical 98.0 - 100.5 %
NF - Assay (KH₂PO₄) (dried basis) 98.0 - 100.5 %
Solubility (5 in 50) Passes Test
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Arsenic (As) ≤3 ppm
NF - Lead (Pb) ≤5 ppm
NF - Insoluble Substances ≤0.2 %
NF - Fluoride (F) ≤10 ppm
NF - Loss on Drying at 105°C ≤1.0 %
NF - Endotoxin Concentration (EU/g) ≤5
NF - pH of a 1 in 100 Solution 4.2 - 4.7
Appearance (colorless to white needle- like crystals)
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR