tri-Natriumcitrat Dihydrat 99,0-100,5% (durch Trockensubstanz), Kristalle, GenAR® USP, Multikompendial, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

7773-06EA 241.07 EUR
7773-06 7773-88 7773-04 7773-26
tri-Natriumcitrat Dihydrat 99,0-100,5% (durch Trockensubstanz), Kristalle, GenAR® USP, Multikompendial, Macron Fine Chemicals™
tri-Natriumcitrat Dihydrat
Formel: Na₃C₆H₅O₇·2H₂O
Schmelzpunkt: 150 °C
Dichte: 1,76 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150031
CAS-Nummer: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets J.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical 99.0 - 100.5 %
USP - Assay (C₆H₅Na₃O₇) (anhydrous basis) 99.0 - 100.5 %
USP - Alkalinity Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Identification C Passes Test
USP - Tartrate Passes Test
USP - Water (H₂O) 10.0 - 13.0 %
EP/BP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis) 99.0 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Acidity or Alkalinity Passes Test
EP/BP - Chloride (Cl) ≤50 ppm
EP/BP - Oxalate (C₂O₄) ≤300 ppm
EP/BP - Sulfate (SO₄) ≤150 ppm
EP/BP - Readily Carbonizable Substances Passes Test
EP/BP - Water (H₂O) 11.0 - 13.0 %
EP/BP - Appearance of Solution Passes Test
Endotoxin Concentration (EU/g) ≤5
JP - Assay (anhydrous basis) 99.0 - 101.0 %
JP - pH (1 in 20) 7.5 - 8.5
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤0.015 %
JP - Identification Passes Test
JP - Sulfate (SO₄) ≤0.048 %
JP - Heavy Metals (as Pb) ≤10 ppm
JP - Arsenic (As) ≤2 ppm
JP - Tartrate Passes Test
JP - Oxalate (C₂O₄) Passes Test
JP - Readily Carbonizable Substances Passes Test
JP - Loss on Drying at 180°C 10.0 - 13.0 %
Appearance (fine, white to slightly yellow crystals) 10.0 - 13.0 %
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR