ortho-Borsäure ≥99,5%, Granulat, GenAR® NF, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

Synonyme: Borsäure, Borhydroxid

Veränderungsmanagement-Kategorie= R

7779-06EA 317.69 EUR
7779-06
ortho-Borsäure ≥99,5%, Granulat, GenAR® NF, Macron Fine Chemicals™
ortho-Borsäure
Formel: B(OH)₃
Molecular Weight: 61,83 g/mol
Siedepunkt: 185 °C (1013 hPa)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011337
CAS-Nummer: 10043-35-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets N.F. Requirements
Meets J.P. Chemical Specifications
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical 99.5 - 100.5 %
NF - Assay (H₃BO₃) (dried basis) 99.5 - 100.5 %
NF - Completeness of Solution Passes Test
NF - Identification Passes Test
NF - Loss on Drying ≤0.5 %
NF - Solubility in Alcohol Passes Test
NF - LAL Bacterial Endotoxins (EU/g) ≤35
EP/BP - Assay (H₃BO₃) 99.0 - 100.5 %
EP/BP - Appearance of Solution Passes Test
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - pH of a 3.3% Solution 3.8 - 4.8
EP/BP- Organic Matter Passes Test
EP/BP - Solubility in Alcohol Passes Test
EP/BP - Sulfate (SO₄) ≤450 ppm
JP - Assay (H₃BO₃) (dried basis) 99.5 - 101.0 %
JP - Identification Passes Test
JP - Clarity and Color of Solution Passes Test
JP - Heavy Metals (as Pb) ≤10 ppm
JP - Arsenic (As) ≤5 ppm
JP - Loss on Drying ≤0.5 %
No Class 1,2,3 or other solvents are used or produced in the = 0.5 %
manufacturing or purification of the product.
Not for Internal Use
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR