(S)-(-)-1-Phenylethylisocyanat, Supelco®
Lieferant: Merck
Synonyme:
(S)-(-)-α-Methylbenzylisocyanat, (S)-(-)-1-Phenethylisocyanat, (S)-(-)-α-Phenethylisocyanat
M-Clarity™ Qualitätsstufe = MQ100
77970-1ML
77970-1MLEA
161
EUR
77970-1ML
(S)-(-)-1-Phenylethylisocyanat, Supelco®
(S)-(-)-1-Phenylethylisocyanat
Formel:
C₉H₉NO Molecular Weight: 147,18 g/mol Siedepunkt: 55 °C (1013 hPa) Dichte: 1,05 g/cm³ (20 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00002036 CAS-Nummer: 14649-03-7 EINECS: 238-698-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...