Thallium(I)ethanolat 96%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Thallium(I)ethoxid, Thallium(I)ethylat, Ethanol Thallium(I)salz
78131.06
78131.06EA
144
EUR
78131.06
Thallium(I)ethanolat 96%
Thallium(I)ethanolat
Purity: 98%
CAS No.: 20398-06-5
Molecular Formula: C2H5OTl
Formula Weight: 249.43
Flash Point:
Formel:
C₂H₅OTl Molecular Weight: 249.44 g/mol Siedepunkt: 130 °C (Zers.) Schmelzpunkt: –3 °C Dichte: 3.493 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009072 CAS-Nummer: 20398-06-5 EINECS: 243-786-2 UN: 2810 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...