Natriumnitrit ≥97,0%, körnig, AR® ACS, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

7824-20EA 1393.43 EUR
7824-20
Natriumnitrit ≥97,0%, körnig, AR® ACS, Macron Fine Chemicals™
Sodium nitrite

Granular.

Formel: NaNO₂
Siedepunkt: 320 °C (1013 hPa)
Schmelzpunkt: 284 °C
Dichte: 2,168 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011118
CAS-Nummer: 7632-00-0
EINECS: 231-555-9
UN: 1500
ADR: 5.1,III
Merck Index: 13,08721

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (NaNO₂) ≥97.0 %
ACS - Insoluble Matter ≤0.01 %
ACS - Chloride (Cl) ≤0.005 %
ACS - Sulfate (SO₄) ≤0.01 %
ACS - Heavy Metals (as Pb) ≤0.001 %
ACS - Iron (Fe) ≤0.001 %
ACS - Calcium (Ca) ≤0.01 %
ACS - Potassium (K) ≤0.005 %
Appearance (white or yellowish white crystalline granules)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR