N-Methyl-bis-heptafluorobutyramide (MBHFBA) ≥96,0% (durch GC), LiChropur™ zur GC-Derivatisierung, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

78268-1ML-F
78268-1ML-FEA 168 EUR
78268-1ML-F
N-Methyl-bis-heptafluorobutyramide (MBHFBA) ≥96,0% (durch GC), LiChropur™ zur GC-Derivatisierung, Supelco®
N-Methyl-bis-heptafluorobutyramide (MBHFBA)
Formel: C₉H₃F₁₄NO₂
Molecular Weight: 423,10 g/mol
MDL: MFCD00049280
CAS-Nummer: 73980-71-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18d4dacb0cd3c7d0a7baf9a0865796a3dad6a0c5&vid=136e5eed639e37865d0fb371b3b202186b174c26&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR