Halt™ Protease and Phosphatase Inhibitor Cocktail
Lieferant: Thermo Fisher Scientific
Halt protease and phosphatase inhibitor cocktail provides the convenience of a single solution with full sample protection. This broad-spectrum inhibitor cocktail is specifically optimised to protect proteins from degradation during extraction and purification. The cocktail contains inhibitors against the major classes of proteases and phosphatases, targeting aminopeptidases, cysteine and serine proteases and serine/threonine and protein tyrosine phosphatases.
- Convenient all-in-one cocktail contains both protease and phosphatase inhibitors
- Immediate inhibition - no need to thaw, open multiple tubes or wait for tablets to dissolve
- Compatible with Thermo Scientific™ Pierce™ cell lysis reagents
- Compatible with mass spectrometry (MS)