Natriumdodecylsulfat (SDS) 20% in wässriger Lösung
Lieferant: G Bioscienes
786-017
786-017EA
422
EUR
786-017
Natriumdodecylsulfat (SDS) 20% in wässriger Lösung
Natriumdodecylsulfat (SDS)
SDS is an anionic detergent molecule tightly bind with the protein molecule masking their native charge and rendering the protein molecules with an overall negative charge.
- Complete disruption and denaturation of protein structure
- Used for solubilisation of proteins including membrane proteins
Formel:
CH₃(CH₂)₁₁OSO₃Na |
MDL:
MFCD00036175 CAS-Nummer: 151-21-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...