Phosphate buffered saline (PBS), 10X
Lieferant: G Bioscienes
786-027
786-027EA
90
EUR
786-027
Phosphate buffered saline (PBS), 10X
Puffer
Elektrophorese-Puffer
Waschpuffer
PBS 10X solution is used as a wash buffers for Western blotting, ELISA and other applications.