TCEP-HCl (Tris(2-carboxyethyl)phosphin Hydrochlorid)

Lieferant: G Bioscienes
Gefahr

Synonyme: TCEP HCl

786-030
786-030EA 196 EUR
786-030
TCEP-HCl (Tris(2-carboxyethyl)phosphin Hydrochlorid)
TCEP-HCl (Tris(2-carboxyethyl)phosphin Hydrochlorid)
Formel: C₉H₁₆ClO₆P
Molecular Weight: 286.65 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00145469
CAS-Nummer: 51805-45-9
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR