Bestatin Hydrochlorid ([S-(R*,S*)]-N-(3-Amino-2-hydroxy-4-phenylbutyroyl)-L-leucin Hydrochlorid)

Lieferant: G Bioscienes

Synonyme: Bestatin hydrochloride, (S)-2-((2S,3R)-3-Amino-2-hydroxy-4-phenylbutanamido)-4-methylpentanoic acid hydrochloride, Bestatin HCl

786-047
786-047EA 668 EUR
786-047
Bestatin Hydrochlorid ([S-(R*,S*)]-N-(3-Amino-2-hydroxy-4-phenylbutyroyl)-L-leucin Hydrochlorid)
Bestatin Hydrochlorid ([S-(R*,S*)]-N-(3-Amino-2-hydroxy-4-phenylbutyroyl)-L-leucin Hydrochlorid)

Bestatin.HCl is a competitive inhibitor of surface aminopeptidases, including aminopeptidase B (Ki=2nM), leucine aminopeptidase (Ki=20nM). Also inhibits aminopeptidases N; does not inhibit endoproteases. Solubility: Soluble up to 5 mg/ml in methanol or 1 mg/ml in 0,15 M NaCl.

Formel: C₁₆H₂₅ClN₂O₄
Molecular Weight: 308.4 · 36.5 g/mol
MDL: MFCD00058004
CAS-Nummer: 65391-42-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR