Pepstatin A
Lieferant: G Bioscienes
786-052
786-052EA 492 EUR
786-052
Pepstatin A
Proteine und Peptide
Pepstatin is a potent inhibitor of various aspartic proteases, including cathepsin D, renin, pepsin, bacterial aspartic proteases and HIV proteases. Soluble in DMSO or methanol.