AEBSF Hydrochlorid (4-(2-Aminoethyl)benzolsulfonylfluorid Hydrochlorid)
Lieferant: G Bioscienes
Synonyme:
4-(2-Aminoethyl)benzenesulphonyl fluoride hydrochloride, AEBSF hydrochloride
786-053
786-053EA
2000
EUR
786-053
AEBSF Hydrochlorid (4-(2-Aminoethyl)benzolsulfonylfluorid Hydrochlorid)
AEBSF Hydrochlorid (4-(2-Aminoethyl)benzolsulfonylfluorid Hydrochlorid)
AEBSF (4-[2-Aminoethyl] benzenesulfonyl fluoride hydrochloride) is a specific irreversible inhibitor of serine proteases, including chymotrypsin, kallikrein, plasmin, thrombin and trypsin. AEBSF is a stable non-toxic alternative to PMSF.
Formel:
C₈H₁₁ClFNO₂S Molecular Weight: 239,69 g/mol |
MDL:
MFCD00132962 CAS-Nummer: 30827-99-7 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...