ALLN
Lieferant: G Bioscienes
786-057
786-057EA 566 EUR
786-057
ALLN
Proteine und Peptide
ALLN is a cell permeable peptide aldehyde inhibitor of calpain I and to a lesser extent calpain II. It also inhibits other neutral cysteine proteases, cathepsin B and L and the proteasome.