RNase-DETECT™
Lieferant: G Bioscienes
786-115 786-116
786-115EA 475 EUR
786-115 786-116
RNase-DETECT™
Tests Nukleinsäure-Tests
RNase-DETECT™ is a sensitive method to detect RNase contamination.
  • Contains aliquots of calibrated RNase substrate for detection of femtogram levels of RNase contamination
  • Serial Dilution Ladder (SDL) detection technique supplied for rapid, clear detection of contamination
  • Compatible with a wide variety of reagent solutions, including SDS, Tris, salts, agarose
  • Suitable to assay solutions that can not be treated with DEPC
Lieferumfang: Supplied with RNase substrate vials, reaction-loading dyes, and protocol for quick analysis of the results.