Wasser, DEPC-behandelt

Lieferant: G Bioscienes

786-109 786-117 786-118
786-109EA 161 EUR
786-109 786-117 786-118
Wasser, DEPC-behandelt
Wasser

Ultra pure DEPC-Treated water is suitable for use with sensitive RNA research. It is prepared by incubating with 0.1% DEPC (diethylpyrocarbonate), an RNase inhibitor, and then autoclaved to remove the DEPC, to prevent downstream interference, and sterilized.

Formel: H₂O
MDL: MFCD00011332
CAS-Nummer: 7732-18-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR