GET™ Total RNA kit
Lieferant: G Bioscienes
786-132
786-132EA
631
EUR
786-132
GET™ Total RNA kit
Nukleinsäure-Reagenzien
Reinigungskits und Reagenzien für Nukleinsäure
The GET™ Total RNA Kit isolates total RNA from contaminating DNA, proteins, and nucleases using our GET™ RNA spin columns. After homogenisation, RNA is bound to the GET™ RNA spin columns and washed. The eluted RNA is ready for any procedure including Northern/slot/dot blots, reverse transcription or RNase protective assays.
- Rapidly inhibit destructive RNases
- Strong chaotropic buffer for rapid RNA release
- High affinity RNA binding spin columns
- Optional DNase treatment
- Compatible with numerous species and tissues