Blocking solution, BLOK™ BSA
Lieferant: G Bioscienes
786-193
786-193EA 393 EUR
786-193
Blocking solution, BLOK™ BSA
Puffer Elektrophorese-Puffer Blockier-Puffer
BLOK™ BSA is a 10% BSA protein blocking solution.
  • For blocking Westerns, ELISA and dot blots
  • Ready-to-use
  • Available in TBS or PBS buffers