Blocking solution, BLOK™ BSA
Lieferant: G Bioscienes
786-193
786-193EA
393
EUR
786-193
Blocking solution, BLOK™ BSA
Puffer
Elektrophorese-Puffer
Blockier-Puffer
BLOK™ BSA is a 10% BSA protein blocking solution.
- For blocking Westerns, ELISA and dot blots
- Ready-to-use
- Available in TBS or PBS buffers