CytoScan™ LDH Cytotoxicity Assay kit
Lieferant: G Bioscienes
CytoScan™ LDH Cytotoxicity assay quantitatively measures the cytosolic, lactate dehydrogenase (LDH) enzyme, which is released from damaged cells. The released LDH is measured with a coupled enzymatic reaction that results in the conversion of a tetrazolium salt (iodonitrotetrazolium; (INT)) into a red color formazan by diaphorase. The resulting formazan is measured at 490 nm.
- Colorimetric assay
- Quantitatively measures LDH release
- For the detection of cell toxicity, death, viability or proliferation
- Ideal for high throughput screening