CytoScan™ LDH Cytotoxicity Assay kit
Lieferant: G Bioscienes
786-210
786-210EA 901 EUR
786-210
CytoScan™ LDH Cytotoxicity Assay kit
Tests Zelltests
CytoScan™ LDH Cytotoxicity assay quantitatively measures the cytosolic, lactate dehydrogenase (LDH) enzyme, which is released from damaged cells. The released LDH is measured with a coupled enzymatic reaction that results in the conversion of a tetrazolium salt (iodonitrotetrazolium; (INT)) into a red color formazan by diaphorase. The resulting formazan is measured at 490 nm.
  • Colorimetric assay
  • Quantitatively measures LDH release
  • For the detection of cell toxicity, death, viability or proliferation
  • Ideal for high throughput screening