Extraction buffers, FOCUS™
Lieferant: G Bioscienes
786-219 786-220 786-221 786-222 786-223 786-233 786-234
786-219EA 348 EUR
786-219 786-220 786-221 786-222 786-223 786-233 786-234
Extraction buffers, FOCUS™
Puffer Extraktionspuffer
FOCUS™ Extraction Buffers are experimentally optimised concentrations of critical agents, buffering and stabilising agents, including urea, thiourea, Nonidet® P-40, CHAPS, and sulphobetaines (SB). They are designed to produce optimal protein extraction and improved spot resolution for 2D gel analysis.