Extraction buffers, FOCUS™
Lieferant: G Bioscienes
786-219
786-220
786-221
786-222
786-223
786-233
786-234
786-219EA
348
EUR
786-219
786-220
786-221
786-222
786-223
786-233
786-234
Extraction buffers, FOCUS™
Puffer
Extraktionspuffer
FOCUS™ Extraction Buffers are experimentally optimised concentrations of critical agents, buffering and stabilising agents, including urea, thiourea, Nonidet® P-40, CHAPS, and sulphobetaines (SB). They are designed to produce optimal protein extraction and improved spot resolution for 2D gel analysis.