Equilibration buffer for IPG strips
Lieferant: G Bioscienes
786-224
786-224EA
204
EUR
786-224
Equilibration buffer for IPG strips
Puffer
Biologische Puffer
Add an appropriate volume of the supplied rehydration buffer to the dry, ready-to-use, urea based equilibration buffer.