Agarose Sealing Solution
Lieferant: G-Biosciences
786-226
786-226EA 160 EUR
786-226
Agarose Sealing Solution
Elektrophorese-Systeme
Simply heat and use for sealing the IPG strips for SDS-PAGE analysis. The Agarose Sealing Solution is prepared in a proprietary buffer to minimise reoxidation of the competing thiol pairs as proteins enter into the second dimension gel. Improves resolution and prevents streaking of protein spots on 2D gels.